The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018242.2 | NP_060712.2 | 10 - 11 | 1007 | 64 |
GenBank mRNA | AK222625.1 | - | 10 - 11 | 1011 | 64 |
AK293196.1 | - | 3 - 4 | 300 | 64 | |
AK301854.1 | - | 9 - 10 | 964 | 64 | |
AK302620.1 | - | 9 - 10 | 909 | 64 | |
AK304647.1 | - | 2 - 3 | 186 | 64 | |
AK310270.1 | - | 8 - 9 | 686 | 64 | |
BC010661.2 | - | 10 - 11 | 996 | 64 | |
BC050592.1 | - | 10 - 11 | 981 | 64 |