Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC47A1
Entrez Gene ID
55244
Gene Name
solute carrier family 47 member 1
Gene Aliases
MATE1
Chromosome Location
Chr.17: 19533854 - 19579033 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018242.2 NP_060712.2 15 - 16 1490 109
GenBank mRNA AK001709.1 - 15 - 16 1490 109
AK298254.1 - 6 - 7 732 109
AK304647.1 - 7 - 8 669 109
BC010661.2 - 15 - 16 1479 109
BC050592.1 - 15 - 16 1464 109
BC058882.1 - 12 - 13 1366 109

View Full Product Details