Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SLC47A1
Entrez Gene ID
55244
Gene Name
solute carrier family 47 member 1
Gene Aliases
MATE1
Chromosome Location
Chr.17: 19533854 - 19579033 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_018242.2 NP_060712.2 5 - 6 584 101
GenBank mRNA AK001709.1 - 5 - 6 584 101
AK222625.1 - 5 - 6 588 101
AK301854.1 - 4 - 5 541 101
BC010661.2 - 5 - 6 573 101
BC050592.1 - 5 - 6 558 101
BC058882.1 - 5 - 6 560 101

View Full Product Details