Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AKNA
Entrez Gene ID
80709
Gene Name
AT-hook transcription factor
Gene Aliases
-
Chromosome Location
Chr.9: 114334149 - 114398498 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001317950.1 NP_001304879.1 16 - 17 3610 165
NM_001317952.1 NP_001304881.1 15 - 16 3168 165
NM_030767.5 NP_110394.3 16 - 17 3555 165
XM_005252244.2 XP_005252301.1 18 - 19 3727 165
XM_005252245.1 XP_005252302.1 17 - 18 3585 165
XM_005252247.4 XP_005252304.1 16 - 17 3606 165
XM_005252248.2 XP_005252305.1 16 - 17 3723 165
XM_006717294.1 XP_006717357.1 16 - 17 3539 165
XM_011519063.2 XP_011517365.1 15 - 16 3218 165
XM_011519064.2 XP_011517366.1 16 - 17 3723 165
XM_011519065.2 XP_011517367.2 16 - 17 3198 165
XM_017015172.1 XP_016870661.1 17 - 18 3325 165
GenBank mRNA AF286341.1 - 6 - 7 1110 165
AK024431.1 - 14 - 15 2503 165
AK074040.1 - 15 - 16 2598 165
AK131082.2 - 16 - 17 3610 165
AK160382.1 - 16 - 17 3612 165
AL833220.1 - 13 - 14 2032 165
AY703039.1 - 17 - 18 3210 165
AY703040.1 - 15 - 16 3168 165
AY703043.1 - 15 - 16 3528 165
AY703044.1 - 14 - 15 3222 165
AY703045.1 - 7 - 8 1305 165
AY703046.1 - 3 - 4 431 165
BC042202.1 - 2 - 3 285 165
BC055285.1 - 16 - 17 3555 165

View Full Product Details