The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001320032.1 | NP_001306961.1 | 20 - 21 | 3247 | 77 |
NM_005688.3 | NP_005679.2 | 20 - 21 | 3132 | 77 | |
XM_005247058.4 | XP_005247115.1 | 20 - 21 | 4700 | 77 | |
XM_005247059.4 | XP_005247116.1 | 20 - 21 | 3122 | 77 | |
XM_011512314.1 | XP_011510616.1 | 20 - 21 | 3023 | 77 | |
XM_011512315.1 | XP_011510617.1 | 20 - 21 | 3182 | 77 | |
XM_017005492.1 | XP_016860981.1 | 17 - 18 | 4371 | 77 | |
GenBank mRNA | AB005659.1 | - | 14 - 15 | 2209 | 77 |
AB019002.1 | - | 19 - 20 | 2998 | 77 | |
AB209103.1 | - | 17 - 18 | 2640 | 77 | |
AB209454.1 | - | 20 - 21 | 3059 | 77 | |
AF104942.1 | - | 20 - 21 | 3071 | 77 | |
AF146074.1 | - | 20 - 21 | 3142 | 77 | |
AY196484.1 | - | 19 - 20 | 2958 | 77 | |
BC140771.1 | - | 19 - 20 | 3008 | 77 | |
U83661.2 | - | 20 - 21 | 3065 | 77 |