The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135095.1 | NP_001128567.1 | 12 - 13 | 1483 | 111 |
NM_022763.3 | NP_073600.3 | 12 - 13 | 1556 | 111 | |
XM_017007062.1 | XP_016862551.1 | 11 - 12 | 1254 | 111 | |
XM_017007063.1 | XP_016862552.1 | 7 - 8 | 740 | 111 | |
XM_017007064.1 | XP_016862553.1 | 12 - 13 | 1536 | 111 | |
GenBank mRNA | AB098597.1 | - | 11 - 12 | 1441 | 111 |
AK075220.1 | - | 2 - 3 | 111 | ||
AK127826.1 | - | 12 - 13 | 111 | ||
AK223599.1 | - | 12 - 13 | 111 | ||
AK314478.1 | - | 12 - 13 | 1483 | 111 | |
AY358146.1 | - | 5 - 6 | 111 | ||
AY358367.1 | - | 12 - 13 | 111 | ||
BC039297.1 | - | 12 - 13 | 111 | ||
BX648340.1 | - | 12 - 13 | 1478 | 111 |