The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135095.1 | NP_001128567.1 | 18 - 19 | 2172 | 63 |
NM_022763.3 | NP_073600.3 | 18 - 19 | 2245 | 63 | |
XM_017007062.1 | XP_016862551.1 | 17 - 18 | 1943 | 63 | |
XM_017007063.1 | XP_016862552.1 | 13 - 14 | 1429 | 63 | |
GenBank mRNA | AB098597.1 | - | 17 - 18 | 2130 | 63 |
AK027052.1 | - | 2 - 3 | 228 | 63 | |
AK075220.1 | - | 8 - 9 | 867 | 63 | |
AK127826.1 | - | 18 - 19 | 2225 | 63 | |
AK223599.1 | - | 18 - 19 | 2276 | 63 | |
AK314478.1 | - | 18 - 19 | 2172 | 63 | |
AY358146.1 | - | 11 - 12 | 1118 | 63 | |
BC033635.2 | - | 6 - 7 | 720 | 63 | |
BC039297.1 | - | 18 - 19 | 2136 | 63 | |
BX648340.1 | - | 18 - 19 | 2167 | 63 |