The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001135095.1 | NP_001128567.1 | 21 - 22 | 2619 | 80 |
NM_022763.3 | NP_073600.3 | 21 - 22 | 2692 | 80 | |
XM_017007062.1 | XP_016862551.1 | 20 - 21 | 2390 | 80 | |
XM_017007063.1 | XP_016862552.1 | 16 - 17 | 1876 | 80 | |
GenBank mRNA | AB098597.1 | - | 20 - 21 | 2577 | 80 |
AK027052.1 | - | 5 - 6 | 675 | 80 | |
AK075220.1 | - | 11 - 12 | 1314 | 80 | |
AK127826.1 | - | 21 - 22 | 2672 | 80 | |
AK223599.1 | - | 21 - 22 | 2723 | 80 | |
AK314478.1 | - | 21 - 22 | 2619 | 80 | |
AL157482.1 | - | 3 - 4 | 340 | 80 | |
AY358146.1 | - | 14 - 15 | 1565 | 80 | |
BC033635.2 | - | 9 - 10 | 1167 | 80 | |
BX648340.1 | - | 21 - 22 | 2614 | 80 |