Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
C1orf21
Entrez Gene ID
81563
Gene Name
chromosome 1 open reading frame 21
Gene Aliases
PIG13
Chromosome Location
Chr.1: 184387016 - 184629021 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_030806.3 NP_110433.1 3 - 4 668 94
GenBank mRNA AF312864.1 - 3 - 4 590 94
AK056487.1 - 3 - 4 404 94
AK312062.1 - 2 - 3 305 94
AY258286.1 - 3 - 4 581 94
BC028567.2 - 3 - 4 626 94

View Full Product Details