Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
BFSP2
Entrez Gene ID
8419
Gene Name
beaded filament structural protein 2
Gene Aliases
CP47, CP49, CTRCT12, LIFL-L, PHAKOSIN
Chromosome Location
Chr.3: 133397882 - 133475212 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_003571.3 NP_003562.1 3 - 4 819 79
XM_017007315.1 XP_016862804.1 3 - 4 2932 79
GenBank mRNA BC113518.1 - 3 - 4 805 79
BC113520.1 - 3 - 4 805 79
U48224.1 - 3 - 4 730 79

View Full Product Details