The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013339.3 | NP_037471.2 | 9 - 10 | 1119 | 72 |
GenBank mRNA | AF063604.2 | - | 9 - 10 | 1074 | 72 |
AF102851.1 | - | 9 - 10 | 1052 | 72 | |
AK022700.1 | - | 9 - 10 | 1057 | 72 | |
AK295286.1 | - | 3 - 4 | 365 | 72 | |
BC001253.1 | - | 9 - 10 | 1051 | 72 | |
CB215801.1 | - | 1 - 2 | 45 | 72 |