The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077262.1 | NP_001070730.1 | 7 - 8 | 763 | 153 |
NM_145345.2 | NP_663320.2 | 11 - 12 | 1243 | 153 | |
NM_183008.2 | NP_892120.2 | 12 - 13 | 1248 | 153 | |
GenBank mRNA | AF370425.1 | - | 1 - 2 | 1036 | 152 |
AF520989.1 | - | 10 - 11 | 952 | 153 | |
AF521017.1 | - | 10 - 11 | 953 | 153 | |
AF521018.1 | - | 10 - 11 | 940 | 153 | |
AK090792.1 | - | 6 - 7 | 1722 | 153 | |
AK093258.1 | - | 12 - 13 | 1192 | 153 | |
AK094198.1 | - | 7 - 8 | 695 | 153 | |
AK097368.1 | - | 11 - 12 | 1341 | 153 | |
AY007163.1 | - | 8 - 9 | 707 | 153 | |
BC038106.1 | - | 10 - 11 | 2041 | 153 | |
BX647774.1 | - | 10 - 11 | 2037 | 153 | |
BX648412.1 | - | 9 - 10 | 1938 | 153 |