The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077262.1 | NP_001070730.1 | 6 - 7 | 639 | 84 |
NM_145345.2 | NP_663320.2 | 10 - 11 | 1119 | 84 | |
NM_183008.2 | NP_892120.2 | 11 - 12 | 1124 | 84 | |
GenBank mRNA | AF520989.1 | - | 9 - 10 | 828 | 84 |
AF521017.1 | - | 9 - 10 | 829 | 84 | |
AF521018.1 | - | 9 - 10 | 816 | 84 | |
AK093258.1 | - | 11 - 12 | 1068 | 84 | |
AK094198.1 | - | 6 - 7 | 571 | 84 | |
AK097368.1 | - | 10 - 11 | 1217 | 84 | |
AY007163.1 | - | 7 - 8 | 583 | 84 |