Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AGPAT3
- Entrez Gene ID
-
56894
- Gene Name
- 1-acylglycerol-3-phosphate O-acyltransferase 3
- Gene Aliases
-
LPAAT-GAMMA1, LPAAT3
- Chromosome Location
-
Chr.21: 43865160 - 43987594 on Build GRCh38
View Full Product Details