Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
PIGT
Entrez Gene ID
51604
Gene Name
phosphatidylinositol glycan anchor biosynthesis class T
Gene Aliases
CGI-06, MCAHS3, NDAP, PNH2
Chromosome Location
Chr.20: 45416067 - 45426245 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001184728.2 NP_001171657.1 5 - 6 696 91
NM_001184729.2 NP_001171658.1 6 - 7 864 91
NM_001184730.2 NP_001171659.1 4 - 5 558 91
NM_015937.5 NP_057021.2 6 - 7 864 91
NR_047691.1 6 - 7 824 91
NR_047692.1 5 - 6 767 91
NR_047693.1 5 - 6 763 91
NR_047694.1 5 - 6 686 91
NR_047695.1 3 - 4 457 91
XM_005260430.2 XP_005260487.1 4 - 5 536 91
XM_005260432.2 XP_005260489.1 5 - 6 658 91
GenBank mRNA AB057724.1 - 6 - 7 774 91
AF132940.1 - 6 - 7 768 91
AK075469.1 - 6 - 7 788 91
AK225517.1 - 6 - 7 794 91
AK293294.1 - 5 - 6 687 91
AK293686.1 - 5 - 6 609 91
AK296063.1 - 5 - 6 693 91
AK296139.1 - 5 - 6 619 91
AK297830.1 - 1 - 2 323 91
AK299013.1 - 3 - 4 383 91
AK301815.1 - 5 - 6 661 91
AK302093.1 - 4 - 5 516 91
AK311100.1 - 5 - 6 646 91
AK311416.1 - 4 - 5 508 91
AK311492.1 - 6 - 7 759 91
AK311520.1 - 4 - 5 560 91
AK311522.1 - 5 - 6 675 91
AK316358.1 - 6 - 7 750 91
AL121742.1 - 6 - 7 767 91
AY358588.1 - 6 - 7 771 91
BC015022.1 - 6 - 7 793 91
BC110892.1 - 6 - 7 772 91
BC136827.1 - 6 - 7 802 91
BC136828.1 - 6 - 7 802 91
BX537612.1 - 4 - 5 2202 91

View Full Product Details