Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRIM29
Entrez Gene ID
23650
Gene Name
tripartite motif containing 29
Gene Aliases
ATDC
Chromosome Location
Chr.11: 120111286 - 120138179 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF230389.1 - 10 - 11 2025 66

View Full Product Details