The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012096.2 | NP_036228.1 | 12 - 13 | 1248 | 139 |
XM_011533583.2 | XP_011531885.1 | 13 - 14 | 1301 | 139 | |
GenBank mRNA | AB037849.2 | - | 12 - 13 | 1101 | 139 |
AF169797.1 | - | 12 - 13 | 1159 | 139 | |
AF424738.1 | - | 12 - 13 | 1157 | 139 | |
AK299008.1 | - | 13 - 14 | 1290 | 139 | |
AK310716.1 | - | 12 - 13 | 1196 | 139 | |
BC028599.2 | - | 12 - 13 | 1248 | 139 |