The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_012096.2 | NP_036228.1 | 18 - 19 | 1841 | 151 |
XM_011533583.2 | XP_011531885.1 | 19 - 20 | 1894 | 151 | |
GenBank mRNA | AB037849.2 | - | 18 - 19 | 1694 | 151 |
AF169797.1 | - | 18 - 19 | 1752 | 151 | |
AF424738.1 | - | 18 - 19 | 1750 | 151 | |
AK299008.1 | - | 19 - 20 | 1883 | 151 | |
AK310716.1 | - | 18 - 19 | 1789 | 151 | |
BC028599.2 | - | 18 - 19 | 1841 | 151 |