Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CATIP
Entrez Gene ID
375307
Gene Name
ciliogenesis associated TTC17 interacting protein
Gene Aliases
C2orf62
Chromosome Location
Chr.2: 218356757 - 218368393 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320865.1 NP_001307794.1 5 - 6 522 103
NM_198559.1 NP_940961.1 5 - 6 489 103
XM_005246539.4 XP_005246596.1 4 - 5 486 103
XM_005246541.4 XP_005246598.1 3 - 4 285 103
XM_011511149.2 XP_011509451.1 3 - 4 284 103
XM_011511150.2 XP_011509452.1 3 - 4 250 103
GenBank mRNA BC052750.1 - 5 - 6 489 103

View Full Product Details