Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FAP
Entrez Gene ID
2191
Gene Name
fibroblast activation protein alpha
Gene Aliases
DPPIV, FAPA, SIMP
Chromosome Location
Chr.2: 162170684 - 162243535 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001291807.1 NP_001278736.1 13 - 14 1352 137
NM_004460.3 NP_004451.2 14 - 15 1427 137
XM_011510796.2 XP_011509098.1 13 - 14 1507 137
XM_011510797.2 XP_011509099.1 14 - 15 1368 137
XM_017003585.1 XP_016859074.1 14 - 15 1368 137
GenBank mRNA AK297118.1 - 13 - 14 1275 137
AK310293.1 - 2 - 3 456 137
AK315448.1 - 14 - 15 1297 137
BC026250.1 - 14 - 15 1260 137
KU257678.1 - 12 - 13 1138 137
U09278.1 - 14 - 15 1427 137
U76833.1 - 14 - 15 1267 137

View Full Product Details