The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001098209.1 | NP_001091679.1 | 12 - 13 | 2221 | 76 |
NM_001098210.1 | NP_001091680.1 | 12 - 13 | 2221 | 76 | |
NM_001904.3 | NP_001895.1 | 12 - 13 | 2221 | 76 | |
XM_005264886.2 | XP_005264943.1 | 12 - 13 | 2235 | 76 | |
XM_006712983.1 | XP_006713046.1 | 12 - 13 | 2133 | 76 | |
XM_006712984.1 | XP_006713047.1 | 12 - 13 | 2133 | 76 | |
XM_006712985.1 | XP_006713048.1 | 12 - 13 | 2195 | 76 | |
XM_017005738.1 | XP_016861227.1 | 12 - 13 | 2235 | 76 | |
GenBank mRNA | AB062292.1 | - | 7 - 8 | 1184 | 76 |
AB451264.1 | - | 10 - 11 | 1953 | 76 | |
AB451392.1 | - | 10 - 11 | 1953 | 76 | |
AK289932.1 | - | 13 - 14 | 2109 | 76 | |
AK294778.1 | - | 13 - 14 | 2334 | 76 | |
AK296790.1 | - | 12 - 13 | 2041 | 76 | |
AK299954.1 | - | 12 - 13 | 1903 | 76 | |
AK314776.1 | - | 12 - 13 | 2063 | 76 | |
BC058926.1 | - | 12 - 13 | 2163 | 76 | |
X87838.1 | - | 12 - 13 | 2167 | 76 | |
Z19054.1 | - | 12 - 13 | 2153 | 76 |