The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_003054.4 | NP_003045.2 | 8 - 9 | 1002 | 99 |
GenBank mRNA | AK314997.1 | - | 8 - 9 | 955 | 99 |
BC030593.1 | - | 7 - 8 | 1260 | 99 | |
BC108927.1 | - | 7 - 8 | 867 | 99 | |
BC108928.2 | - | 7 - 8 | 867 | 99 | |
L09118.1 | - | 8 - 9 | 944 | 99 | |
L14269.1 | - | 8 - 9 | 872 | 99 | |
L23205.1 | - | 8 - 9 | 952 | 99 | |
X71354.1 | - | 8 - 9 | 902 | 99 |