Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
GNG7
Entrez Gene ID
2788
Gene Name
G protein subunit gamma 7
Gene Aliases
-
Chromosome Location
Chr.19: 2511219 - 2702748 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_052847.2 NP_443079.1 2 - 3 165 116
XM_017026606.1 XP_016882095.1 2 - 3 274 116
GenBank mRNA AK311750.1 - 2 - 3 102 116
BC014466.1 - 2 - 3 126 116
BC053630.1 - 2 - 3 82 116
DA106844.1 - 2 - 3 165 116
DA247608.1 - 2 - 3 109 116

View Full Product Details