Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
Chromosome Location
Chr.11: 85957171 - 86069881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008660.2 NP_001008660.1 1 - 2 444 94
NM_001206946.1 NP_001193875.1 1 - 2 444 94
NM_007166.3 NP_009097.2 1 - 2 444 94
XM_005274322.2 XP_005274379.1 1 - 2 428 94
XM_005274323.2 XP_005274380.1 1 - 2 428 94
XM_005274324.2 XP_005274381.1 1 - 2 428 94
XM_005274325.2 XP_005274382.1 1 - 2 428 94
XM_005274326.2 XP_005274383.1 1 - 2 428 94
XM_005274327.2 XP_005274384.1 1 - 2 428 94
XM_005274328.2 XP_005274385.1 1 - 2 428 94
XM_005274329.3 XP_005274386.1 1 - 2 428 94
XM_005274330.2 XP_005274387.1 1 - 2 428 94
XM_005274331.2 XP_005274388.1 1 - 2 428 94
XM_005274332.2 XP_005274389.1 1 - 2 428 94
XM_005274333.2 XP_005274390.1 1 - 2 428 94
XM_005274334.2 XP_005274391.1 1 - 2 428 94
XM_005274335.2 XP_005274392.1 1 - 2 428 94
XM_005274336.2 XP_005274393.1 1 - 2 428 94
XM_005274337.2 XP_005274394.1 1 - 2 428 94
XM_005274340.2 XP_005274397.1 1 - 2 428 94
XM_006718699.2 XP_006718762.1 1 - 2 428 94
XM_006718700.2 XP_006718763.1 1 - 2 428 94
XM_006718701.3 XP_006718764.1 1 - 2 428 94
XM_011545293.1 XP_011543595.1 1 - 2 428 94
XM_017018381.1 XP_016873870.1 1 - 2 428 94
XM_017018382.1 XP_016873871.1 1 - 2 428 94
XM_017018383.1 XP_016873872.1 1 - 2 428 94
XM_017018384.1 XP_016873873.1 1 - 2 428 94
XM_017018385.1 XP_016873874.1 1 - 2 428 94
XM_017018386.1 XP_016873875.1 1 - 2 428 94
XM_017018387.1 XP_016873876.1 1 - 2 428 94
XM_017018388.1 XP_016873877.1 1 - 2 428 94
GenBank mRNA AB210017.1 - 1 - 2 444 94
AB451308.1 - 1 - 2 127 94
AK291414.1 - 2 - 3 311 94
BC048259.1 - 1 - 2 205 94
BC064357.1 - 1 - 2 406 94
BC073961.1 - 1 - 2 280 94
U45976.1 - 1 - 2 274 94

View Full Product Details