Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
PICALM
Entrez Gene ID
8301
Gene Name
phosphatidylinositol binding clathrin assembly protein
Gene Aliases
CALM, CLTH, LAP
Chromosome Location
Chr.11: 85957171 - 86069881 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008660.2 NP_001008660.1 12 - 13 1571 97
NM_001206947.1 NP_001193876.1 12 - 13 1286 97
XM_005274330.2 XP_005274387.1 12 - 13 1555 97
XM_005274332.2 XP_005274389.1 12 - 13 1555 97
XM_005274333.2 XP_005274390.1 12 - 13 1555 97
XM_005274335.2 XP_005274392.1 12 - 13 1555 97
XM_005274336.2 XP_005274393.1 12 - 13 1555 97
XM_005274340.2 XP_005274397.1 12 - 13 1555 97
XM_017018382.1 XP_016873871.1 12 - 13 1555 97
XM_017018383.1 XP_016873872.1 12 - 13 1555 97
XM_017018384.1 XP_016873873.1 12 - 13 1555 97
XM_017018386.1 XP_016873875.1 12 - 13 1555 97
XM_017018387.1 XP_016873876.1 12 - 13 1555 97
GenBank mRNA AB451308.1 - 12 - 13 1254 97
AK297062.1 - 4 - 5 456 97
AK300275.1 - 12 - 13 1263 97
BC064357.1 - 12 - 13 1533 97
BC073961.1 - 12 - 13 1407 97

View Full Product Details