Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
JAG2
Entrez Gene ID
3714
Gene Name
jagged 2
Gene Aliases
HJ2, SER2
Chromosome Location
Chr.14: 105140981 - 105168824 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_002226.4 NP_002217.3 4 - 5 1130 64
NM_145159.2 NP_660142.1 4 - 5 1130 64
GenBank mRNA AF003521.1 - 4 - 5 758 64
AF020201.1 - 4 - 5 777 64
AF029778.1 - 4 - 5 1130 64
AF029779.1 - 4 - 5 1130 64
BC032053.1 - 4 - 5 793 64
Y14330.1 - 4 - 5 678 64

View Full Product Details