Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CARS
Entrez Gene ID
833
Gene Name
cysteinyl-tRNA synthetase
Gene Aliases
CARS1, CYSRS, MGC:11246
Chromosome Location
Chr.11: 3000922 - 3057451 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001014437.2 NP_001014437.1 12 - 13 1480 88
NM_001194997.1 NP_001181926.1 12 - 13 1480 88
NM_001751.5 NP_001742.1 11 - 12 1231 88
NM_139273.3 NP_644802.1 11 - 12 1231 88
NR_036542.1 12 - 13 1527 88
XM_006718341.3 XP_006718404.1 11 - 12 1254 88
XM_017018389.1 XP_016873878.1 11 - 12 1254 88
XM_017018390.1 XP_016873879.1 12 - 13 1494 88
XM_017018391.1 XP_016873880.1 9 - 10 1478 88
XM_017018392.1 XP_016873881.1 11 - 12 1416 88
GenBank mRNA AB208814.1 - 10 - 11 3405 88
AF288206.1 - 11 - 12 1164 88
AF288207.1 - 11 - 12 1164 88
AK096313.1 - 12 - 13 1494 88
AK222614.1 - 11 - 12 1172 88
AK289432.1 - 11 - 12 1204 88
AK295371.1 - 11 - 12 1122 88
AK296763.1 - 12 - 13 1345 88
AK298514.1 - 12 - 13 1447 88
AK302644.1 - 12 - 13 1447 88
AK308005.1 - 12 - 13 1400 88
BC002880.2 - 11 - 12 1217 88
BT009913.1 - 11 - 12 1147 88
BX647906.1 - 12 - 13 1454 88
L06845.1 - 9 - 10 1018 88

View Full Product Details