The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001014437.2 | NP_001014437.1 | 7 - 8 | 886 | 71 |
NM_001194997.1 | NP_001181926.1 | 7 - 8 | 886 | 71 | |
NM_001751.5 | NP_001742.1 | 6 - 7 | 637 | 71 | |
NM_139273.3 | NP_644802.1 | 6 - 7 | 637 | 71 | |
NR_036542.1 | 7 - 8 | 933 | 71 | ||
XM_006718341.3 | XP_006718404.1 | 6 - 7 | 660 | 71 | |
XM_017018389.1 | XP_016873878.1 | 6 - 7 | 660 | 71 | |
XM_017018390.1 | XP_016873879.1 | 7 - 8 | 900 | 71 | |
XM_017018391.1 | XP_016873880.1 | 4 - 5 | 884 | 71 | |
XM_017018392.1 | XP_016873881.1 | 6 - 7 | 822 | 71 | |
GenBank mRNA | AB208814.1 | - | 6 - 7 | 613 | 71 |
AF288206.1 | - | 6 - 7 | 570 | 71 | |
AF288207.1 | - | 6 - 7 | 570 | 71 | |
AK096313.1 | - | 7 - 8 | 900 | 71 | |
AK222614.1 | - | 6 - 7 | 578 | 71 | |
AK289432.1 | - | 6 - 7 | 610 | 71 | |
AK295371.1 | - | 6 - 7 | 528 | 71 | |
AK296763.1 | - | 7 - 8 | 751 | 71 | |
AK298514.1 | - | 7 - 8 | 853 | 71 | |
AK302644.1 | - | 7 - 8 | 853 | 71 | |
AK308005.1 | - | 7 - 8 | 806 | 71 | |
BC002880.2 | - | 6 - 7 | 623 | 71 | |
BT009913.1 | - | 6 - 7 | 553 | 71 | |
BX647906.1 | - | 7 - 8 | 860 | 71 | |
L06845.1 | - | 4 - 5 | 424 | 71 |