The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256702.1 | NP_001243631.1 | 6 - 7 | 739 | 91 |
NM_006537.3 | NP_006528.2 | 7 - 8 | 871 | 91 | |
NR_046341.1 | 8 - 9 | 1024 | 91 | ||
NR_046342.1 | 8 - 9 | 987 | 91 | ||
XM_017022763.1 | XP_016878252.1 | 9 - 10 | 1870 | 91 | |
XM_017022764.1 | XP_016878253.1 | 8 - 9 | 1806 | 91 | |
XM_017022765.1 | XP_016878254.1 | 8 - 9 | 1012 | 91 | |
XM_017022766.1 | XP_016878255.1 | 9 - 10 | 1055 | 91 | |
XM_017022767.1 | XP_016878256.1 | 10 - 11 | 1986 | 91 | |
XM_017022768.1 | XP_016878257.1 | 10 - 11 | 2060 | 91 | |
GenBank mRNA | AF073344.1 | - | 7 - 8 | 735 | 91 |
AF077040.1 | - | 6 - 7 | 515 | 91 | |
AK022931.1 | - | 5 - 6 | 458 | 91 | |
AK301236.1 | - | 6 - 7 | 655 | 91 | |
AK307810.1 | - | 7 - 8 | 630 | 91 | |
AK309382.1 | - | 6 - 7 | 692 | 91 | |
AK314223.1 | - | 7 - 8 | 790 | 91 | |
AY461579.1 | - | 8 - 9 | 900 | 91 | |
BC018113.2 | - | 7 - 8 | 736 | 91 | |
BC065300.1 | - | 7 - 8 | 771 | 91 | |
BC100029.1 | - | 7 - 8 | 743 | 91 | |
BC107137.1 | - | 7 - 8 | 731 | 91 | |
BC107138.1 | - | 7 - 8 | 731 | 91 | |
BT007269.1 | - | 7 - 8 | 642 | 91 |