The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001256702.1 | NP_001243631.1 | 8 - 9 | 1002 | 68 |
NM_006537.3 | NP_006528.2 | 9 - 10 | 1134 | 68 | |
NR_046341.1 | 10 - 11 | 1287 | 68 | ||
NR_046342.1 | 10 - 11 | 1250 | 68 | ||
XM_017022763.1 | XP_016878252.1 | 11 - 12 | 2133 | 68 | |
XM_017022764.1 | XP_016878253.1 | 10 - 11 | 2069 | 68 | |
XM_017022765.1 | XP_016878254.1 | 10 - 11 | 1275 | 68 | |
XM_017022766.1 | XP_016878255.1 | 11 - 12 | 1318 | 68 | |
XM_017022767.1 | XP_016878256.1 | 12 - 13 | 2249 | 68 | |
XM_017022768.1 | XP_016878257.1 | 12 - 13 | 2323 | 68 | |
GenBank mRNA | AF073344.1 | - | 9 - 10 | 69 | |
AF077040.1 | - | 8 - 9 | 778 | 68 | |
AK022931.1 | - | 7 - 8 | 721 | 68 | |
AK300948.1 | - | 7 - 8 | 792 | 68 | |
AK301236.1 | - | 8 - 9 | 918 | 68 | |
AK301563.1 | - | 3 - 4 | 433 | 68 | |
AK307810.1 | - | 9 - 10 | 893 | 68 | |
AK309382.1 | - | 8 - 9 | 955 | 68 | |
AK314223.1 | - | 9 - 10 | 1053 | 68 | |
AY461579.1 | - | 10 - 11 | 1163 | 68 | |
BC018113.2 | - | 9 - 10 | 999 | 68 | |
BC065300.1 | - | 9 - 10 | 1034 | 68 | |
BC100029.1 | - | 9 - 10 | 1006 | 68 | |
BC107137.1 | - | 9 - 10 | 994 | 68 | |
BC107138.1 | - | 9 - 10 | 994 | 68 | |
BT007269.1 | - | 9 - 10 | 905 | 68 | |
BX647760.1 | - | 10 - 11 | 1028 | 68 |