Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EYA1
Entrez Gene ID
2138
Gene Name
EYA transcriptional coactivator and phosphatase 1
Gene Aliases
BOP, BOR, BOS1, OFC1
Chromosome Location
Chr.8: 71197433 - 71548130 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000503.5 NP_000494.2 16 - 17 2236 156
NM_001288574.1 NP_001275503.1 16 - 17 2218 156
NM_001288575.1 NP_001275504.1 16 - 17 2154 156
NM_172058.3 NP_742055.1 15 - 16 1828 156
NM_172059.3 NP_742056.1 14 - 15 1545 156
NM_172060.3 NP_742057.1 14 - 15 1700 156
XM_011517483.2 XP_011515785.1 15 - 16 6533 156
XM_011517484.2 XP_011515786.2 17 - 18 1979 156
XM_017013201.1 XP_016868690.1 17 - 18 2022 156
XM_017013202.1 XP_016868691.1 17 - 18 1866 156
XM_017013203.1 XP_016868692.1 17 - 18 1991 156
XM_017013204.1 XP_016868693.1 17 - 18 1977 156
XM_017013205.1 XP_016868694.1 17 - 18 1994 156
XM_017013206.1 XP_016868695.1 18 - 19 1975 156
XM_017013207.1 XP_016868696.1 16 - 17 1903 156
XM_017013208.1 XP_016868697.1 16 - 17 2258 156
XM_017013209.1 XP_016868698.1 16 - 17 1778 156
XM_017013210.1 XP_016868699.1 16 - 17 1887 156
XM_017013211.1 XP_016868700.1 16 - 17 1866 156
XM_017013212.1 XP_016868701.1 15 - 16 2133 156
XM_017013213.1 XP_016868702.1 15 - 16 6671 156
GenBank mRNA AF467247.1 - 14 - 15 1520 156
AJ000097.1 - 15 - 16 1773 156
AJ000098.1 - 16 - 17 2234 156
AK127798.1 - 16 - 17 1654 156
AK295897.1 - 16 - 17 1727 156
BC121798.1 - 14 - 15 2059 156
BC121799.1 - 15 - 16 1740 156
DB004805.1 - 5 - 6 527 156

View Full Product Details