Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC38
Entrez Gene ID
55020
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
LL22NC03-5H6.5
Chromosome Location
Chr.22: 46267964 - 46294008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_017931.2 NP_060401.2 7 - 8 811 75
XM_011530259.1 XP_011528561.1 8 - 9 75
GenBank mRNA AK000706.1 - 7 - 8 767 75
AK300606.1 - 6 - 7 617 75
BC018918.2 - 7 - 8 752 75

View Full Product Details