Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZC3H13
Entrez Gene ID
23091
Gene Name
zinc finger CCCH-type containing 13
Gene Aliases
KIAA0853
Chromosome Location
Chr.13: 45954469 - 46052785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_005266301.3 XP_005266358.1 18 - 19 5125 154
XM_005266302.3 XP_005266359.1 18 - 19 4825 154
XM_005266303.3 XP_005266360.1 19 - 20 5150 154
XM_005266304.3 XP_005266361.1 18 - 19 5107 154
XM_005266305.3 XP_005266362.1 19 - 20 5143 154
XM_005266306.3 XP_005266363.1 18 - 19 4821 154
XM_005266307.3 XP_005266364.1 18 - 19 5122 154
XM_005266308.3 XP_005266365.1 17 - 18 5041 154
XM_005266309.3 XP_005266366.1 17 - 18 4741 154
XM_005266310.3 XP_005266367.1 17 - 18 5023 154
XM_005266311.3 XP_005266368.1 17 - 18 5038 154
XM_017020475.1 XP_016875964.1 19 - 20 4913 154
XM_017020476.1 XP_016875965.1 19 - 20 4910 154
XM_017020477.1 XP_016875966.1 18 - 19 5066 154
XM_017020478.1 XP_016875967.1 18 - 19 5059 154
XM_017020479.1 XP_016875968.1 18 - 19 4829 154
XM_017020480.1 XP_016875969.1 17 - 18 4737 154
XM_017020481.1 XP_016875970.1 18 - 19 5063 154
XM_017020482.1 XP_016875971.1 17 - 18 4738 154
GenBank mRNA AB020660.1 - 6 - 7 2570 154
AK001815.1 - 3 - 4 432 154
AK074924.1 - 4 - 5 1798 154

View Full Product Details