Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- ZC3H13
- Entrez Gene ID
-
23091
- Gene Name
- zinc finger CCCH-type containing 13
- Gene Aliases
-
KIAA0853
- Chromosome Location
-
Chr.13: 45954469 - 46052785 on Build GRCh38
View Full Product Details