The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030630.2 | NP_085133.1 | 18 - 19 | 2452 | 75 |
XM_005257226.1 | XP_005257283.1 | 18 - 19 | 2447 | 75 | |
GenBank mRNA | AK074401.1 | - | 18 - 19 | 2380 | 75 |
AK075111.1 | - | 8 - 9 | 977 | 75 | |
AK125514.1 | - | 5 - 6 | 1273 | 75 | |
AK304081.1 | - | 18 - 19 | 2336 | 75 | |
BC032219.1 | - | 14 - 15 | 1750 | 75 | |
BC035372.1 | - | 18 - 19 | 2368 | 75 |