Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TTC38
Entrez Gene ID
55020
Gene Name
tetratricopeptide repeat domain 38
Gene Aliases
LL22NC03-5H6.5
Chromosome Location
Chr.22: 46267964 - 46294008 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq XM_011530259.1 XP_011528561.1 3 - 4 309 78

View Full Product Details