The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001297595.1 | NP_001284524.1 | 14 - 15 | 2623 | 69 |
NM_001297597.1 | NP_001284526.1 | 5 - 6 | 1729 | 69 | |
NM_015260.3 | NP_056075.1 | 15 - 16 | 2719 | 69 | |
XM_006722704.1 | XP_006722767.1 | 5 - 6 | 1334 | 69 | |
GenBank mRNA | AB014600.2 | - | 14 - 15 | 2610 | 69 |
AK001633.1 | - | 8 - 9 | 1724 | 69 | |
AK295618.1 | - | 5 - 6 | 1668 | 69 | |
AY706204.1 | - | 15 - 16 | 2699 | 69 | |
BC005113.1 | - | 2 - 3 | 461 | 69 |