Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146115.1 NP_001139587.1 13 - 14 1763 70
NM_001146116.1 NP_001139588.1 14 - 15 1880 70
NM_015151.3 NP_055966.2 14 - 15 1892 70
NM_206889.2 NP_996772.1 14 - 15 1892 70
NM_206890.2 NP_996773.1 14 - 15 1892 70
NM_206891.2 NP_996774.1 14 - 15 1892 70
XM_011529492.2 XP_011527794.1 15 - 16 2007 70
XM_011529501.1 XP_011527803.1 15 - 16 2007 70
XM_011529502.1 XP_011527804.1 15 - 16 2007 70
XM_017028291.1 XP_016883780.1 15 - 16 2007 70
XM_017028292.1 XP_016883781.1 15 - 16 2007 70
XM_017028293.1 XP_016883782.1 15 - 16 2004 70
XM_017028294.1 XP_016883783.1 14 - 15 1886 70
XM_017028295.1 XP_016883784.1 14 - 15 1883 70
XM_017028296.1 XP_016883785.1 14 - 15 1883 70
XM_017028297.1 XP_016883786.1 14 - 15 1875 70
XM_017028298.1 XP_016883787.1 13 - 14 1754 70
XM_017028299.1 XP_016883788.1 14 - 15 1752 70
XM_017028300.1 XP_016883789.1 15 - 16 2007 70
XM_017028301.1 XP_016883790.1 15 - 16 2007 70
XM_017028302.1 XP_016883791.1 13 - 14 1647 70
XM_017028303.1 XP_016883792.1 15 - 16 2007 70
XM_017028304.1 XP_016883793.1 15 - 16 2007 70
XM_017028305.1 XP_016883794.1 15 - 16 2007 70
GenBank mRNA AB273729.1 - 14 - 15 1730 70
AF432263.1 - 9 - 10 1102 70
AF490768.1 - 14 - 15 1785 70
AK056738.1 - 14 - 15 1784 70
AK098236.1 - 9 - 10 1035 70
AK303351.1 - 12 - 13 1707 70
BC033718.1 - 13 - 14 1746 70
BC038443.1 - 14 - 15 1891 70

View Full Product Details