Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146115.1 NP_001139587.1 19 - 20 2449 55
NM_001146116.1 NP_001139588.1 20 - 21 2566 55
NM_015151.3 NP_055966.2 20 - 21 2578 55
NM_206889.2 NP_996772.1 20 - 21 2578 55
NM_206890.2 NP_996773.1 20 - 21 2578 55
XM_011529492.2 XP_011527794.1 21 - 22 2693 55
XM_011529501.1 XP_011527803.1 21 - 22 2693 55
XM_011529502.1 XP_011527804.1 21 - 22 2693 55
XM_017028291.1 XP_016883780.1 21 - 22 2693 55
XM_017028292.1 XP_016883781.1 21 - 22 2693 55
XM_017028293.1 XP_016883782.1 21 - 22 2690 55
XM_017028294.1 XP_016883783.1 20 - 21 2572 55
XM_017028295.1 XP_016883784.1 20 - 21 2569 55
XM_017028296.1 XP_016883785.1 20 - 21 2569 55
XM_017028297.1 XP_016883786.1 20 - 21 2561 55
XM_017028298.1 XP_016883787.1 19 - 20 2440 55
XM_017028299.1 XP_016883788.1 20 - 21 2438 55
XM_017028300.1 XP_016883789.1 21 - 22 2693 55
XM_017028301.1 XP_016883790.1 21 - 22 2693 55
XM_017028302.1 XP_016883791.1 19 - 20 2333 55
XM_017028303.1 XP_016883792.1 21 - 22 2693 55
XM_017028304.1 XP_016883793.1 21 - 22 2693 55
XM_017028305.1 XP_016883794.1 21 - 22 2693 55
GenBank mRNA AB273729.1 - 20 - 21 2416 55
AF490768.1 - 20 - 21 2471 55
AK056738.1 - 20 - 21 2470 55
AK091790.1 - 4 - 5 1616 55
AK303351.1 - 18 - 19 2393 55
BC033718.1 - 19 - 20 2432 55
BC038443.1 - 20 - 21 2577 55
BQ017399.1 - 1 - 2 55

View Full Product Details