Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146116.1 NP_001139588.1 23 - 24 3005 71
NM_015151.3 NP_055966.2 23 - 24 3017 71
XM_011529492.2 XP_011527794.1 24 - 25 3132 71
XM_011529501.1 XP_011527803.1 24 - 25 3132 71
XM_017028291.1 XP_016883780.1 24 - 25 3132 71
XM_017028292.1 XP_016883781.1 24 - 25 3132 71
XM_017028293.1 XP_016883782.1 24 - 25 3129 71
XM_017028294.1 XP_016883783.1 23 - 24 3011 71
XM_017028295.1 XP_016883784.1 23 - 24 3008 71
XM_017028296.1 XP_016883785.1 23 - 24 3008 71
XM_017028297.1 XP_016883786.1 23 - 24 3000 71
XM_017028298.1 XP_016883787.1 22 - 23 2879 71
XM_017028299.1 XP_016883788.1 23 - 24 2877 71
XM_017028300.1 XP_016883789.1 24 - 25 3132 71
XM_017028301.1 XP_016883790.1 24 - 25 3132 71
XM_017028302.1 XP_016883791.1 22 - 23 2772 71
XM_017028303.1 XP_016883792.1 24 - 25 3132 71
XM_017028304.1 XP_016883793.1 24 - 25 3132 71
XM_017028305.1 XP_016883794.1 24 - 25 3128 71
GenBank mRNA AB273729.1 - 23 - 24 2855 71
AF490768.1 - 23 - 24 2910 71
AK303351.1 - 21 - 22 2832 71

View Full Product Details