Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DIP2A
Entrez Gene ID
23181
Gene Name
disco interacting protein 2 homolog A
Gene Aliases
C21orf106, DIP2
Chromosome Location
Chr.21: 46458942 - 46570900 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001146116.1 NP_001139588.1 34 - 35 4253 65
NM_015151.3 NP_055966.2 34 - 35 4265 65
XM_011529492.2 XP_011527794.1 35 - 36 4380 65
XM_017028291.1 XP_016883780.1 35 - 36 4380 65
XM_017028292.1 XP_016883781.1 35 - 36 4380 65
XM_017028293.1 XP_016883782.1 35 - 36 4377 65
XM_017028294.1 XP_016883783.1 34 - 35 4259 65
XM_017028295.1 XP_016883784.1 34 - 35 4256 65
XM_017028296.1 XP_016883785.1 34 - 35 4256 65
XM_017028297.1 XP_016883786.1 34 - 35 4248 65
XM_017028298.1 XP_016883787.1 33 - 34 4127 65
XM_017028299.1 XP_016883788.1 34 - 35 4125 65
XM_017028302.1 XP_016883791.1 33 - 34 4020 65
XM_017028304.1 XP_016883793.1 35 - 36 5252 65
GenBank mRNA AB273729.1 - 34 - 35 4103 65
AF490768.1 - 34 - 35 4158 65
AK054807.1 - 1 - 2 514 65

View Full Product Details