Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD28
Entrez Gene ID
940
Gene Name
CD28 molecule
Gene Aliases
Tp44
Chromosome Location
Chr.2: 203706475 - 203739756 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001243078.1 NP_001230007.1 1 - 2 273 83
GenBank mRNA AF222342.1 - 1 - 2 51 83

View Full Product Details