Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
EIF4G1
Entrez Gene ID
1981
Gene Name
eukaryotic translation initiation factor 4 gamma 1
Gene Aliases
EIF-4G1, EIF4F, EIF4G, EIF4GI, P220, PARK18
Chromosome Location
Chr.3: 184314495 - 184335358 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AF002815.1 - 1 - 2 215 139
DC347118.1 - 1 - 2 378 139

View Full Product Details