The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NR_024010.1 | 3 - 4 | 995 | 67 | |
GenBank mRNA | AK293381.1 | - | 3 - 4 | 566 | 67 |
BP262099.1 | - | 3 - 4 | 293 | 67 |
Gene Name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromosome | |
---|---|---|---|---|---|---|
Upstream | UDP glucuronosyltransferase family 2 member B10 | UGT2B10 | UDPGT2B10 | -100.6kb | Anti-sense | Chr. - 68815981 - 68832038 on Build GRCh38 |
Downstream | UDP glucuronosyltransferase family 2 member B7 | UGT2B7 | UDPGT 2B9, UDPGT2B7, UDPGTH2, UGT2B9 | 107.9kb | Anti-sense | Chr. - 69051264 - 69112987 on Build GRCh38 |