The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001077706.2 | NP_001071174.1 | 16 - 17 | 2359 | 116 |
NM_001195037.2 | NP_001181966.1 | 15 - 16 | 2219 | 116 | |
XM_006715472.3 | XP_006715535.1 | 17 - 18 | 2478 | 116 | |
XM_011535795.2 | XP_011534097.1 | 16 - 17 | 2349 | 116 | |
XM_011535797.2 | XP_011534099.1 | 14 - 15 | 2061 | 116 | |
XM_017010828.1 | XP_016866317.1 | 17 - 18 | 2451 | 116 | |
XM_017010829.1 | XP_016866318.1 | 16 - 17 | 2385 | 116 | |
GenBank mRNA | BC146884.1 | - | 14 - 15 | 2066 | 116 |
DQ904331.1 | - | 14 - 15 | 2013 | 116 | |
DV080594.1 | - | 1 - 2 | 116 |