Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
SGIP1
Entrez Gene ID
84251
Gene Name
SH3 domain GRB2 like endophilin interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.1: 66533272 - 66751139 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001308203.1 NP_001295132.1 8 - 9 626 74
NM_032291.3 NP_115667.2 10 - 11 908 98
XM_005271264.3 XP_005271321.1 10 - 11 742 98
XM_005271268.3 XP_005271325.1 9 - 10 658 98
XM_005271270.4 XP_005271327.1 8 - 9 965 74
XM_006710961.2 XP_006711024.1 10 - 11 742 98
XM_006710966.2 XP_006711029.1 10 - 11 742 98
XM_006710967.2 XP_006711030.1 10 - 11 730 98
XM_006710969.2 XP_006711032.1 9 - 10 658 98
XM_006710971.2 XP_006711034.1 10 - 11 742 98
XM_006710972.2 XP_006711035.1 10 - 11 742 98
XM_006710973.2 XP_006711036.1 8 - 9 718 74
XM_006710974.2 XP_006711037.1 7 - 8 634 74
XM_011542291.1 XP_011540593.1 10 - 11 905 98
XM_011542292.1 XP_011540594.1 10 - 11 905 98
XM_011542293.1 XP_011540595.1 7 - 8 634 74
XM_017002505.1 XP_016857994.1 10 - 11 730 98
XM_017002506.1 XP_016857995.1 11 - 12 853 98
XM_017002507.1 XP_016857996.1 10 - 11 905 98
XM_017002508.1 XP_016857997.1 11 - 12 851 98
XM_017002509.1 XP_016857998.1 8 - 9 706 74
XM_017002510.1 XP_016857999.1 9 - 10 829 74
XM_017002511.1 XP_016858000.1 11 - 12 853 98
XM_017002512.1 XP_016858001.1 11 - 12 726 98
XM_017002513.1 XP_016858002.1 10 - 11 905 98
XM_017002514.1 XP_016858003.1 9 - 10 658 98
XM_017002515.1 XP_016858004.1 7 - 8 634 74
XM_017002516.1 XP_016858005.1 8 - 9 757 74
XM_017002517.1 XP_016858006.1 11 - 12 852 98
XM_017002518.1 XP_016858007.1 10 - 11 780 98
XM_017002519.1 XP_016858008.1 10 - 11 805 98
XM_017002520.1 XP_016858009.1 8 - 9 646 86
XM_017002521.1 XP_016858010.1 9 - 10 658 98
XM_017002522.1 XP_016858011.1 10 - 11 780 98
XM_017002523.1 XP_016858012.1 10 - 11 804 98
XM_017002524.1 XP_016858013.1 9 - 10 822 74
XM_017002525.1 XP_016858014.1 9 - 10 821 74
XM_017002526.1 XP_016858015.1 8 - 9 706 74
XM_017002527.1 XP_016858016.1 9 - 10 730 74
XM_017002528.1 XP_016858017.1 8 - 9 750 74
XM_017002529.1 XP_016858018.1 10 - 11 742 98
XM_017002530.1 XP_016858019.1 9 - 10 718 86
XM_017002531.1 XP_016858020.1 8 - 9 718 74
XM_017002532.1 XP_016858021.1 8 - 9 646 86
XM_017002533.1 XP_016858022.1 9 - 10 820 74
XM_017002534.1 XP_016858023.1 7 - 8 634 74
XM_017002535.1 XP_016858024.1 8 - 9 748 74
XM_017002536.1 XP_016858025.1 9 - 10 658 98
XM_017002537.1 XP_016858026.1 8 - 9 747 74
GenBank mRNA AB210039.1 - 10 - 11 690 98
AK090697.1 - 2 - 3 183 74
AK298970.1 - 8 - 9 595 98
AK299941.1 - 9 - 10 669 98
AL136561.1 - 10 - 11 582 98
BC040516.1 - 11 - 12 655 98
BX640813.1 - 8 - 9 603 74
CR749541.1 - 8 - 9 820 74

View Full Product Details