Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CRADD
Entrez Gene ID
8738
Gene Name
CASP2 and RIPK1 domain containing adaptor with death domain
Gene Aliases
MRT34, RAIDD
Chromosome Location
Chr.12: 93677342 - 93894840 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001320099.1 NP_001307028.1 2 - 3 613 101
NM_003805.4 NP_003796.1 2 - 3 403 101
GenBank mRNA AB451280.1 - 1 - 2 295 101
AB451412.1 - 1 - 2 295 101
AK314861.1 - 2 - 3 383 101
BC017042.1 - 2 - 3 375 101
BC037905.1 - 2 - 3 617 101
BT009837.1 - 1 - 2 295 101
CR407643.1 - 1 - 2 295 101
HY113325.1 - 2 - 3 404 101
U79115.1 - 2 - 3 395 101
U84388.1 - 2 - 3 332 101

View Full Product Details