The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019885.3 | NP_063938.1 | 2 - 3 | 459 | 76 |
XM_005264433.4 | XP_005264490.1 | 2 - 3 | 423 | 76 | |
GenBank mRNA | AF252297.1 | - | 2 - 3 | 431 | 76 |
AK294933.1 | - | 1 - 2 | 429 | 76 | |
AK295683.1 | - | 1 - 2 | 74 | 76 | |
AK313433.1 | - | 2 - 3 | 434 | 76 | |
BC069443.1 | - | 2 - 3 | 431 | 76 | |
BC109205.2 | - | 2 - 3 | 431 | 76 |