Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CYP26B1
Entrez Gene ID
56603
Gene Name
cytochrome P450 family 26 subfamily B member 1
Gene Aliases
CYP26A2, P450RAI-2, P450RAI2, RHFCA
Chromosome Location
Chr.2: 72129238 - 72147862 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001277742.1 NP_001264671.1 2 - 3 509 64
NM_019885.3 NP_063938.1 3 - 4 734 64
XM_005264433.4 XP_005264490.1 3 - 4 698 64
XM_011532988.1 XP_011531290.1 2 - 3 295 64
GenBank mRNA AF252297.1 - 3 - 4 706 64
AK294814.1 - 2 - 3 481 64
AK294933.1 - 2 - 3 704 64
AK295683.1 - 2 - 3 349 64
AK313433.1 - 3 - 4 709 64
BC069443.1 - 3 - 4 706 64
BC109205.2 - 3 - 4 706 64
CR749631.1 - 2 - 3 358 64
DC308258.1 - 2 - 3 509 64
FJ467289.1 - 2 - 3 481 64

View Full Product Details