The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277742.1 | NP_001264671.1 | 4 - 5 | 956 | 106 |
NM_019885.3 | NP_063938.1 | 5 - 6 | 1181 | 106 | |
XM_005264433.4 | XP_005264490.1 | 5 - 6 | 1145 | 106 | |
XM_011532988.1 | XP_011531290.1 | 4 - 5 | 742 | 106 | |
GenBank mRNA | AF252297.1 | - | 5 - 6 | 1153 | 106 |
AK294814.1 | - | 4 - 5 | 928 | 106 | |
AK294933.1 | - | 4 - 5 | 1151 | 106 | |
AK295683.1 | - | 4 - 5 | 796 | 106 | |
AK313433.1 | - | 5 - 6 | 1156 | 106 | |
BC069443.1 | - | 5 - 6 | 1153 | 106 | |
BC109205.2 | - | 5 - 6 | 1153 | 106 | |
CR749631.1 | - | 4 - 5 | 805 | 106 | |
FJ467289.1 | - | 4 - 5 | 928 | 106 |